Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277995.1 | NP_001264924.1 | 3 - 4 | 367 | 145 | |
GenBank mRNA | BX929314.1 | - | 3 - 4 | 368 | 145 | |
BX935860.2 | - | 3 - 4 | 383 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |