Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936953.3 | XP_004937010.3 | 6 - 7 | 1772 | 61 | |
XM_025153539.1 | XP_025009307.1 | 6 - 7 | 951 | 61 | ||
XM_025153540.1 | XP_025009308.1 | 6 - 7 | 1777 | 61 | ||
XM_025153541.1 | XP_025009309.1 | 6 - 7 | 864 | 61 | ||
XM_025153542.1 | XP_025009310.1 | 6 - 7 | 811 | 61 | ||
GenBank mRNA | CR338985.2 | - | 6 - 7 | 885 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |