Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416415.6 | XP_416415.5 | 3 - 4 | 1073 | 84 | |
GenBank mRNA | CR353321.1 | - | 3 - 4 | 388 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |