Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278096.1 | NP_001265025.1 | 8 - 9 | 919 | 115 | |
XM_015293766.2 | XP_015149252.1 | 8 - 9 | 959 | 115 | ||
GenBank mRNA | BX936136.1 | - | 8 - 9 | 906 | 115 | |
BX950663.1 | - | 8 - 9 | 887 | 115 | ||
CR353583.1 | - | 8 - 9 | 919 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |