Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421099.6 | XP_421099.3 | 2 - 3 | 173 | 71 | |
GenBank mRNA | CR385630.1 | - | 2 - 3 | 173 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |