Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416643.6 | XP_416643.3 | 4 - 5 | 807 | 78 | |
GenBank mRNA | BX934649.2 | - | 3 - 4 | 623 | 78 | |
CR385918.1 | - | 2 - 3 | 265 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |