Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291238.2 | XP_015146724.1 | 5 - 6 | 848 | 130 | |
XM_015291240.2 | XP_015146726.1 | 6 - 7 | 1004 | 130 | ||
XM_015291244.2 | XP_015146730.1 | 6 - 7 | 1285 | 130 | ||
XM_015291253.2 | XP_015146739.1 | 5 - 6 | 819 | 130 | ||
XM_015291255.2 | XP_015146741.1 | 5 - 6 | 911 | 130 | ||
XM_025142204.1 | XP_024997972.1 | 6 - 7 | 943 | 130 | ||
XM_416454.6 | XP_416454.1 | 5 - 6 | 1023 | 130 | ||
GenBank mRNA | CR386658.1 | - | 5 - 6 | 869 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |