Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420183.6 | XP_420183.2 | 3 - 4 | 547 | 85 | |
GenBank mRNA | BX929485.2 | - | 3 - 4 | 545 | 85 | |
BX931941.1 | - | 3 - 4 | 537 | 85 | ||
CR387274.1 | - | 3 - 4 | 544 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |