Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946472.3 | XP_004946529.1 | 4 - 5 | 607 | 94 | |
XM_015295986.2 | XP_015151472.1 | 5 - 6 | 650 | 94 | ||
XM_015295987.2 | XP_015151473.1 | 5 - 6 | 644 | 94 | ||
XM_015295988.2 | XP_015151474.1 | 5 - 6 | 650 | 94 | ||
XM_015295989.2 | XP_015151475.1 | 5 - 6 | 649 | 94 | ||
XM_015295990.2 | XP_015151476.1 | 4 - 5 | 601 | 94 | ||
XM_015295991.2 | XP_015151477.1 | 5 - 6 | 475 | 94 | ||
XM_015295993.2 | XP_015151479.1 | 5 - 6 | 501 | 94 | ||
GenBank mRNA | CR387598.1 | - | 4 - 5 | 384 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |