Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281969.1 | XP_015137455.1 | 5 - 6 | 1024 | 105 | |
GenBank mRNA | BX929749.2 | - | 2 - 3 | 303 | 105 | |
BX932420.1 | - | 1 - 2 | 94 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |