Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252160.1 | NP_001239089.1 | 4 - 5 | 413 | 133 | |
GenBank mRNA | BU398441.1 | - | 4 - 5 | 418 | 133 | |
BX932658.1 | - | 4 - 5 | 418 | 133 | ||
BX932692.1 | - | 4 - 5 | 517 | 133 | ||
BX935910.2 | - | 4 - 5 | 359 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |