Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293898.2 | XP_015149384.1 | 3 - 4 | 881 | 77 | |
GenBank mRNA | BX950474.2 | - | 2 - 3 | 168 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |