Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281999.2 | XP_015137485.1 | 5 - 6 | 623 | 76 | |
XM_015282000.2 | XP_015137486.1 | 5 - 6 | 629 | 76 | ||
GenBank mRNA | CR386138.1 | - | 5 - 6 | 574 | 76 | |
CR386940.1 | - | 5 - 6 | 581 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |