Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006214.2 | NP_001006214.1 | 3 - 4 | 316 | 127 | |
GenBank mRNA | AJ719994.1 | - | 3 - 4 | 316 | 127 | |
CR388669.1 | - | 3 - 4 | 284 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |