Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291303.1 | XP_015146789.1 | 3 - 4 | 427 | 58 | |
XM_015291304.2 | XP_015146790.1 | 3 - 4 | 412 | 58 | ||
XM_015291305.2 | XP_015146791.1 | 3 - 4 | 531 | 58 | ||
GenBank mRNA | CR389892.1 | - | 2 - 3 | 402 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |