Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291303.1 | XP_015146789.1 | 7 - 8 | 747 | 79 | |
XM_015291304.2 | XP_015146790.1 | 7 - 8 | 732 | 79 | ||
XM_015291305.2 | XP_015146791.1 | 7 - 8 | 851 | 79 | ||
GenBank mRNA | CR389892.1 | - | 6 - 7 | 722 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |