Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420876.6 | XP_420876.4 | 2 - 3 | 671 | 85 | |
GenBank mRNA | CR390682.1 | - | 1 - 2 | 320 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |