Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275419.2 | XP_015130905.1 | 4 - 5 | 6339 | 83 | |
XM_415250.6 | XP_415250.4 | 4 - 5 | 6117 | 83 | ||
GenBank mRNA | CR390992.1 | - | 1 - 2 | 404 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |