Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282235.2 | NP_001269164.1 | 9 | 2594 | 119 | |
GenBank mRNA | CR391625.1 | - | 1 | 746 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |