Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416217.5 | XP_416217.3 | 1 - 2 | 216 | 99 | |
GenBank mRNA | CR406879.1 | - | 1 - 2 | 197 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |