Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BATF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025150893.1 | XP_025006661.1 | 1 - 2 | 291 | 77 | |
GenBank mRNA | CR523179.1 | - | 1 - 2 | 245 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |