Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199418.3 | NP_001186347.3 | 3 - 4 | 418 | 151 | |
GenBank mRNA | CR523356.1 | - | 2 - 3 | 310 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |