Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STKLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945936.3 | XP_004945993.1 | 15 - 16 | 2110 | 81 | |
XM_025141407.1 | XP_024997175.1 | 14 - 15 | 1978 | 81 | ||
GenBank mRNA | CR523822.1 | - | 4 - 5 | 483 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |