Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234035.3 | XP_001234036.2 | 4 - 5 | 980 | 63 | |
GenBank mRNA | AF461038.1 | - | 3 - 4 | 692 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |