Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231336.5 | XP_001231337.3 | 3 - 4 | 74 | ||
XM_015292302.2 | XP_015147788.1 | 4 - 5 | 74 | |||
XM_015292303.2 | XP_015147789.1 | 3 - 4 | 74 | |||
XM_015292304.2 | XP_015147790.1 | 2 - 3 | 74 | |||
XM_015292305.2 | XP_015147791.1 | 3 - 4 | 74 | |||
XM_015292306.2 | XP_015147792.1 | 3 - 4 | 74 | |||
XM_025154294.1 | XP_025010062.1 | 3 - 4 | 74 | |||
XM_025154295.1 | XP_025010063.1 | 2 - 3 | 74 | |||
GenBank mRNA | AJ719720.1 | - | 3 - 4 | 697 | 73 | |
BU132661.1 | - | 1 - 2 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |