Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201445.1 | NP_001188374.1 | 4 - 5 | 400 | 101 | |
GenBank mRNA | AJ720235.1 | - | 3 - 4 | 482 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |