Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287145.2 | XP_015142631.1 | 4 - 5 | 5729 | 141 | |
XM_015287153.2 | XP_015142639.1 | 3 - 4 | 5608 | 141 | ||
XM_015287164.1 | XP_015142650.1 | 3 - 4 | 1942 | 141 | ||
XM_416367.6 | XP_416367.3 | 4 - 5 | 5719 | 141 | ||
GenBank mRNA | AY928942.1 | - | 2 - 3 | 4633 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |