Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277548.1 | NP_001264477.1 | 4 - 5 | 976 | 98 | |
XM_015287500.2 | XP_015142986.1 | 5 - 6 | 636 | 98 | ||
GenBank mRNA | BX929287.1 | - | 4 - 5 | 989 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |