Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277623.1 | NP_001264552.1 | 2 - 3 | 145 | 109 | |
NM_001277624.1 | NP_001264553.1 | 2 - 3 | 145 | 109 | ||
GenBank mRNA | BG709792.1 | - | 2 - 3 | 145 | 109 | |
BU136195.1 | - | 2 - 3 | 136 | 109 | ||
BU357038.1 | - | 2 - 3 | 139 | 109 | ||
BX929638.1 | - | 2 - 3 | 141 | 109 | ||
BX932269.1 | - | 2 - 3 | 139 | 109 | ||
BX935499.1 | - | 2 - 3 | 136 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |