Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199557.1 | NP_001186486.1 | 4 - 5 | 745 | 101 | |
GenBank mRNA | BX931979.2 | - | 3 - 4 | 568 | 101 | |
BX932028.2 | - | 4 - 5 | 745 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |