Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282235.2 | NP_001269164.1 | 2 - 3 | 420 | 104 | |
GenBank mRNA | CR388896.1 | - | 2 - 3 | 421 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |