Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936012.3 | XP_004936069.1 | 3 - 4 | 360 | 100 | |
XM_015285549.2 | XP_015141035.1 | 5 - 6 | 693 | 100 | ||
XM_420694.6 | XP_420694.2 | 5 - 6 | 841 | 100 | ||
GenBank mRNA | CR390951.1 | - | 1 - 2 | 78 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |