Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937630.3 | XP_004937687.1 | 3 - 4 | 1880 | 57 | |
XM_004937631.3 | XP_004937688.1 | 3 - 4 | 1193 | 57 | ||
XM_015284046.2 | XP_015139532.1 | 4 - 5 | 1226 | 57 | ||
XM_015284049.2 | XP_015139535.1 | 3 - 4 | 1177 | 57 | ||
XM_015284052.2 | XP_015139538.1 | 3 - 4 | 1175 | 57 | ||
XM_015284057.2 | XP_015139543.1 | 3 - 4 | 1469 | 57 | ||
XM_416147.5 | XP_416147.2 | 3 - 4 | 1317 | 57 | ||
GenBank mRNA | CR391122.1 | - | 2 - 3 | 980 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |