Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257327.1 | NP_001244256.1 | 4 - 5 | 650 | 150 | |
GenBank mRNA | CR385157.1 | - | 4 - 5 | 535 | 150 | |
CR406231.1 | - | 4 - 5 | 650 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |