Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282133.2 | XP_015137619.1 | 3 - 4 | 3846 | 83 | |
XM_015282134.2 | XP_015137620.1 | 3 - 4 | 3853 | 83 | ||
XM_015282135.2 | XP_015137621.1 | 3 - 4 | 3852 | 83 | ||
XM_015282136.2 | XP_015137622.1 | 4 - 5 | 3231 | 83 | ||
XM_015282137.2 | XP_015137623.1 | 4 - 5 | 588 | 83 | ||
XM_015282138.2 | XP_015137624.1 | 4 - 5 | 563 | 83 | ||
XM_015282141.2 | XP_015137627.1 | 4 - 5 | 589 | 83 | ||
XM_025148321.1 | XP_025004089.1 | 3 - 4 | 3882 | 83 | ||
GenBank mRNA | BX934162.2 | - | 4 - 5 | 487 | 83 | |
CR406777.1 | - | 4 - 5 | 555 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |