Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288430.2 | XP_015143916.1 | 20 - 21 | 2717 | 67 | |
XM_025151960.1 | XP_025007728.1 | 19 - 20 | 2609 | 67 | ||
XM_025151961.1 | XP_025007729.1 | 20 - 21 | 2717 | 67 | ||
XM_025151962.1 | XP_025007730.1 | 4 - 5 | 432 | 67 | ||
GenBank mRNA | CR407180.1 | - | 2 - 3 | 234 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |