Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278091.1 | NP_001265020.1 | 6 - 7 | 756 | 103 | |
NM_001278092.1 | NP_001265021.1 | 6 - 7 | 698 | 103 | ||
GenBank mRNA | BX935160.1 | - | 6 - 7 | 690 | 103 | |
CR522886.1 | - | 6 - 7 | 588 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |