Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943647.3 | XP_004943704.3 | 10 - 11 | 1195 | 96 | |
XM_015279061.2 | XP_015134547.2 | 9 - 10 | 1132 | 96 | ||
XM_015279062.2 | XP_015134548.2 | 9 - 10 | 1129 | 96 | ||
XM_015279063.2 | XP_015134549.1 | 10 - 11 | 1083 | 96 | ||
XM_015279064.2 | XP_015134550.1 | 9 - 10 | 1026 | 96 | ||
XM_015279065.2 | XP_015134551.1 | 9 - 10 | 1024 | 96 | ||
XM_015279066.2 | XP_015134552.2 | 10 - 11 | 1192 | 96 | ||
XM_025153988.1 | XP_025009756.1 | 9 - 10 | 935 | 96 | ||
XM_425051.6 | XP_425051.5 | 10 - 11 | 1192 | 96 | ||
GenBank mRNA | EF520347.1 | - | 10 - 11 | 1069 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |