Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167718.1 | NP_001161190.1 | 4 - 5 | 953 | 152 | |
NM_001167719.1 | NP_001161191.1 | 4 - 5 | 524 | 152 | ||
GenBank mRNA | M64990.1 | - | 4 - 5 | 1049 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |