Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297854.2 | XP_015153340.1 | 2 - 3 | 1524 | 69 | |
XM_427615.6 | XP_427615.3 | 1 - 2 | 905 | 69 | ||
GenBank mRNA | M85043.1 | - | 2 - 3 | 244 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |