Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415241.6 | XP_415241.4 | 2 - 3 | 322 | 99 | |
GenBank mRNA | BX930077.1 | - | 1 - 2 | 98 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |