Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154966.1 | XP_025010734.1 | 2 - 3 | 95 | 109 | |
XM_414505.6 | XP_414505.3 | 2 - 3 | 95 | 109 | ||
GenBank mRNA | BX930082.1 | - | 2 - 3 | 84 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |