Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941776.3 | XP_004941833.1 | 5 - 6 | 1872 | 114 | |
XM_004941778.3 | XP_004941835.1 | 5 - 6 | 1873 | 114 | ||
XM_015287662.2 | XP_015143148.1 | 5 - 6 | 1873 | 114 | ||
GenBank mRNA | BX930174.1 | - | 1 - 2 | 231 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |