Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941776.3 | XP_004941833.1 | 6 - 7 | 113 | ||
XM_004941778.3 | XP_004941835.1 | 6 - 7 | 113 | |||
XM_015287662.2 | XP_015143148.1 | 6 - 7 | 113 | |||
GenBank mRNA | BX930174.1 | - | 2 - 3 | 336 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |