Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302180.1 | NP_001289109.1 | 1 - 2 | 62 | 66 | |
GenBank mRNA | BU296850.1 | - | 1 - 2 | 62 | 66 | |
BX930499.1 | - | 1 - 2 | 62 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |