Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277583.1 | NP_001264512.1 | 3 - 4 | 321 | 98 | |
XM_015291008.2 | XP_015146494.1 | 4 - 5 | 98 | |||
GenBank mRNA | BX931765.1 | - | 2 - 3 | 240 | 98 | |
CR353506.1 | - | 3 - 4 | 321 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |