Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422738.6 | XP_422738.1 | 1 - 2 | 87 | 74 | |
GenBank mRNA | BX932504.1 | - | 1 - 2 | 61 | 74 | |
BX932869.1 | - | 1 - 2 | 62 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |