Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421207.6 | XP_421207.3 | 3 - 4 | 512 | 134 | |
GenBank mRNA | BX932678.1 | - | 3 - 4 | 513 | 134 | |
CR353992.2 | - | 3 - 4 | 569 | 134 | ||
CR354308.1 | - | 3 - 4 | 534 | 134 | ||
CR405723.1 | - | 3 - 4 | 571 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |