Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151244.1 | XP_025007012.1 | 1 - 2 | 397 | 59 | |
XM_421364.6 | XP_421364.2 | 1 - 2 | 100 | 59 | ||
GenBank mRNA | BX932711.1 | - | 1 - 2 | 82 | 59 | |
CR353802.1 | - | 1 - 2 | 119 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |