Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297000.2 | XP_015152486.1 | 3 - 4 | 366 | 61 | |
XM_015297001.2 | XP_015152487.1 | 4 - 5 | 803 | 61 | ||
XM_025142735.1 | XP_024998503.1 | 3 - 4 | 323 | 61 | ||
GenBank mRNA | BX933771.1 | - | 4 - 5 | 452 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |